The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037738.2 | NP_001032827.1 | 7 - 8 | 821 | 173 |
NM_002520.6 | NP_002511.1 | 7 - 8 | 821 | 173 | |
XM_011534564.1 | XP_011532866.1 | 6 - 7 | 641 | 173 | |
GenBank mRNA | AB042278.1 | - | 7 - 8 | 576 | 173 |
AB451236.1 | - | 7 - 8 | 576 | 173 | |
AB451361.1 | - | 7 - 8 | 576 | 173 | |
AK000472.1 | - | 5 - 6 | 384 | 173 | |
AK290652.1 | - | 7 - 8 | 684 | 173 | |
AY347529.1 | - | 8 - 9 | 711 | 173 | |
AY740634.1 | - | 7 - 8 | 648 | 173 | |
AY740635.1 | - | 7 - 8 | 648 | 173 | |
AY740636.1 | - | 7 - 8 | 648 | 173 | |
AY740637.1 | - | 7 - 8 | 648 | 173 | |
AY740638.1 | - | 7 - 8 | 648 | 173 | |
AY740639.1 | - | 7 - 8 | 648 | 173 | |
AY740640.1 | - | 7 - 8 | 645 | 170 | |
BC002398.2 | - | 7 - 8 | 663 | 173 | |
BC003670.1 | - | 1 - 1 | 378 | 173 | |
BC008495.1 | - | 7 - 8 | 673 | 173 | |
BC012566.1 | - | 7 - 8 | 676 | 173 | |
BC014349.1 | - | 7 - 8 | 672 | 173 | |
BC016716.1 | - | 7 - 8 | 675 | 173 | |
BC016768.1 | - | 7 - 8 | 675 | 173 | |
BC016824.1 | - | 7 - 8 | 675 | 173 | |
BC020467.1 | - | 7 - 8 | 662 | 173 | |
BC021668.1 | - | 7 - 8 | 676 | 173 | |
BC021983.1 | - | 7 - 8 | 674 | 173 | |
BC050628.1 | - | 7 - 8 | 662 | 173 | |
BC107754.1 | - | 7 - 8 | 677 | 173 | |
BT007011.1 | - | 7 - 8 | 576 | 173 | |
DQ303464.1 | - | 7 - 8 | 672 | 173 | |
EF429251.1 | - | 7 - 8 | 576 | 173 | |
KU178237.1 | - | 7 - 8 | 576 | 173 | |
M23613.1 | - | 7 - 8 | 669 | 173 | |
M26697.1 | - | 7 - 8 | 666 | 173 | |
M28699.1 | - | 7 - 8 | 663 | 173 | |
X16934.1 | - | 7 - 8 | 536 | 173 |