The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019024.2 | NP_061897.1 | 30 - 31 | 5003 | 119 |
XM_006712034.2 | XP_006712097.1 | 30 - 31 | 4942 | 119 | |
XM_006712035.3 | XP_006712098.1 | 30 - 31 | 5006 | 119 | |
XM_011532934.2 | XP_011531236.1 | 30 - 31 | 5470 | 119 | |
XM_011532935.2 | XP_011531237.1 | 19 - 20 | 3142 | 119 | |
XM_017004378.1 | XP_016859867.1 | 30 - 31 | 4942 | 119 | |
XM_017004379.1 | XP_016859868.1 | 31 - 32 | 5380 | 119 | |
XM_017004380.1 | XP_016859869.1 | 31 - 32 | 5153 | 119 | |
GenBank mRNA | AB037835.1 | - | 21 - 22 | 3450 | 119 |
AK001513.1 | - | 4 - 5 | 710 | 119 | |
AK026928.1 | - | 7 - 8 | 1263 | 119 | |
AL832880.1 | - | 3 - 4 | 405 | 119 | |
BC150605.1 | - | 30 - 31 | 4988 | 119 | |
BX538008.1 | - | 24 - 25 | 3926 | 119 | |
BX648805.1 | - | 16 - 17 | 3163 | 119 |