Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC4A2
Entrez Gene ID
6522
Gene Name
solute carrier family 4 member 2
Gene Aliases
AE2, BND3L, EPB3L1, HKB3, NBND3
Chromosome Location
Chr.7: 151058212 - 151076527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199692.1 NP_001186621.1 14 - 15 3229 147
NM_001199693.1 NP_001186622.1 13 - 14 2255 147
NM_001199694.1 NP_001186623.1 13 - 14 2676 147
NM_003040.3 NP_003031.3 14 - 15 2445 147
GenBank mRNA AB209158.1 - 14 - 15 3229 147
AK295767.1 - 13 - 14 2255 147
AK312699.1 - 14 - 15 2424 147
BC004893.2 - 14 - 15 2082 147
BC009386.2 - 13 - 14 2676 147
BC009434.1 - 13 - 14 2676 147
BC010069.2 - 14 - 15 2082 147
BC028601.2 - 13 - 14 2277 147
CV807529.1 - 4 - 5 657 147
U62531.1 - 14 - 15 2363 147
X03918.1 - 7 - 8 1061 147
X62137.1 - 14 - 15 2265 147

View Full Product Details