Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1A
Entrez Gene ID
773
Gene Name
calcium voltage-gated channel subunit alpha1 A
Gene Aliases
APCA, BI, CACNL1A4, CAV2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6
Chromosome Location
Chr.19: 13206442 - 13506460 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000068.3 NP_000059.3 24 - 25 4237 144
NM_001127221.1 NP_001120693.1 24 - 25 4228 144
NM_001127222.1 NP_001120694.1 24 - 25 4225 144
NM_001174080.1 NP_001167551.1 24 - 25 4228 144
NM_023035.2 NP_075461.2 24 - 25 4237 144
GenBank mRNA AB035726.1 - 24 - 25 4027 144
AB035727.2 - 24 - 25 3989 144
AF004883.1 - 24 - 25 4237 144
AF004884.1 - 24 - 25 4237 144
FJ040507.1 - 24 - 25 3992 144
U79666.1 - 24 - 25 4209 144
X99897.1 - 24 - 25 4270 144

View Full Product Details