Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PDE4D
- Entrez Gene ID
-
5144
- Gene Name
- phosphodiesterase 4D
- Gene Aliases
-
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
- Chromosome Location
-
Chr.5: 58969038 - 60488098 on Build GRCh38
View Full Product Details