The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032482.2 | NP_115871.1 | 2 - 3 | 158 | 101 |
XM_005259659.3 | XP_005259716.1 | 2 - 3 | 344 | 101 | |
XM_005259660.3 | XP_005259717.1 | 2 - 3 | 344 | 101 | |
XM_011528359.2 | XP_011526661.1 | 2 - 3 | 344 | 101 | |
XM_011528360.1 | XP_011526662.1 | 2 - 3 | 109 | 101 | |
XM_017027366.1 | XP_016882855.1 | 2 - 3 | 344 | 101 | |
GenBank mRNA | AF509504.1 | - | 2 - 3 | 158 | 101 |