The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000170.2 | NP_000161.2 | 10 - 11 | 1588 | 174 |
GenBank mRNA | AK310534.1 | - | 6 - 7 | 963 | 174 |
AK314156.1 | - | 10 - 11 | 1590 | 174 | |
BC111993.1 | - | 10 - 11 | 1461 | 174 | |
BC111995.1 | - | 10 - 11 | 1461 | 174 | |
D90239.1 | - | 10 - 11 | 1545 | 174 | |
M63635.1 | - | 10 - 11 | 1476 | 174 | |
M64590.1 | - | 10 - 11 | 1545 | 174 |