The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199868.1 | NP_001186797.1 | 6 - 7 | 760 | 142 |
NM_001321980.1 | NP_001308909.1 | 7 - 8 | 815 | 142 | |
NM_006541.4 | NP_006532.2 | 6 - 7 | 760 | 142 | |
XM_017015474.1 | XP_016870963.1 | 7 - 8 | 815 | 142 | |
GenBank mRNA | AF118649.1 | - | 6 - 7 | 723 | 142 |
AF118652.1 | - | 6 - 7 | 726 | 142 | |
AJ010841.1 | - | 6 - 7 | 718 | 142 | |
AK022131.1 | - | 6 - 7 | 734 | 142 | |
AK225609.1 | - | 6 - 7 | 723 | 142 | |
BC005289.1 | - | 6 - 7 | 736 | 142 | |
BC014372.1 | - | 5 - 6 | 578 | 142 |