Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM45A
Entrez Gene ID
404636
Gene Name
family with sequence similarity 45 member A
Gene Aliases
-
Chromosome Location
Chr.10: 119104065 - 119137984 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207009.3 NP_996892.1 1 - 2 127 124
NR_130122.1 1 - 2 127 124
XM_017016264.1 XP_016871753.1 1 - 2 108 124
GenBank mRNA AK293365.1 - 1 - 2 104 124
BI753197.1 - 1 - 2 93 124

View Full Product Details