Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNA
Entrez Gene ID
200081
Gene Name
taxilin alpha
Gene Aliases
IL14, TXLN
Chromosome Location
Chr.1: 32179669 - 32198285 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175852.3 NP_787048.1 3 - 4 574 76
XM_017000562.1 XP_016856051.1 2 - 3 867 76
XM_017000564.1 XP_016856053.1 2 - 3 428 76
GenBank mRNA AF516206.1 - 2 - 3 587 76
AL832338.1 - 3 - 4 611 76
AL832636.2 - 3 - 4 633 76
AL832637.2 - 2 - 3 790 76
BC080578.1 - 2 - 3 551 76
BC103823.1 - 2 - 3 539 76
BC103824.1 - 2 - 3 539 76

View Full Product Details