Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PRAP1
- Entrez Gene ID
-
118471
- Gene Name
- proline rich acidic protein 1
- Gene Aliases
-
PRO1195, UPA
- Chromosome Location
-
Chr.10: 133347340 - 133352683 on Build GRCh38
- Gene Symbol
- ZNF511-PRAP1
- Entrez Gene ID
-
104326056
- Gene Name
- ZNF511-PRAP1 readthrough
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EU937518.1 |
- |
7 - 8 |
754 |
149 |
View Full Product Details