Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
Chromosome Location
Chr.12: 49622717 - 49644669 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031698.2 NP_001026868.2 24 - 25 2617 110
NM_012272.2 NP_036404.1 23 - 24 2889 110
XM_006719324.3 XP_006719387.1 23 - 24 3674 110
XM_006719325.3 XP_006719388.1 23 - 24 3671 110
XM_011538135.2 XP_011536437.1 23 - 24 3674 110
XM_011538136.2 XP_011536438.1 23 - 24 3671 110
XM_011538137.2 XP_011536439.1 22 - 23 3593 110
XM_011538138.2 XP_011536440.1 22 - 23 3563 110
XM_011538139.2 XP_011536441.1 24 - 25 2614 110
XM_011538140.2 XP_011536442.1 24 - 25 2611 110
XM_011538141.1 XP_011536443.1 23 - 24 2440 110
XM_011538143.2 XP_011536445.1 23 - 24 2456 110
XM_011538144.2 XP_011536446.1 22 - 23 2329 110
XM_017019135.1 XP_016874624.1 23 - 24 2377 110
XM_017019136.1 XP_016874625.1 22 - 23 2326 110
XM_017019137.1 XP_016874626.1 22 - 23 2326 110
XM_017019139.1 XP_016874628.1 19 - 20 2340 110
GenBank mRNA AK027117.1 - 9 - 10 898 110
AL137459.1 - 10 - 11 1120 110
AL834216.1 - 23 - 24 2889 110
BC031917.1 - 21 - 22 2213 110
BC067364.1 - 23 - 24 2441 110

View Full Product Details