The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322271.1 | NP_001309200.1 | 11 - 12 | 1705 | 119 |
NM_001322272.1 | NP_001309201.1 | 12 - 13 | 2070 | 119 | |
NM_017437.2 | NP_059133.1 | 12 - 13 | 1865 | 119 | |
GenBank mRNA | AB037788.1 | - | 6 - 7 | 989 | 119 |
AK001627.2 | - | 12 - 13 | 1842 | 119 | |
AK001912.1 | - | 10 - 11 | 1421 | 119 | |
AK023583.1 | - | 9 - 10 | 1442 | 119 | |
AL442079.1 | - | 5 - 6 | 764 | 119 | |
BC070095.1 | - | 12 - 13 | 1835 | 119 |