The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322270.1 | NP_001309199.1 | 7 - 8 | 929 | 86 |
NM_001322271.1 | NP_001309200.1 | 6 - 7 | 769 | 86 | |
NM_001322272.1 | NP_001309201.1 | 7 - 8 | 1134 | 86 | |
NM_017437.2 | NP_059133.1 | 7 - 8 | 929 | 86 | |
GenBank mRNA | AB037788.1 | - | 1 - 2 | 53 | 86 |
AK001627.2 | - | 7 - 8 | 906 | 86 | |
AK023583.1 | - | 4 - 5 | 506 | 86 | |
BC070095.1 | - | 7 - 8 | 899 | 86 |