Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESRRB
Entrez Gene ID
2103
Gene Name
estrogen related receptor beta
Gene Aliases
DFNB35, ERR2, ERRb, ESRL2, NR3B2
Chromosome Location
Chr.14: 76371347 - 76501839 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004452.3 NP_004443.3 4 - 5 771 180
XM_011536547.2 XP_011534849.1 2 - 3 933 180
XM_011536550.2 XP_011534852.1 4 - 5 1007 180
XM_011536553.2 XP_011534855.1 2 - 3 933 180
XM_011536554.2 XP_011534856.1 2 - 3 933 180
XM_017021085.1 XP_016876574.1 3 - 4 578 180
XM_017021086.1 XP_016876575.1 4 - 5 1008 180
GenBank mRNA AB307714.1 - 1 - 2 400 180
AY451389.1 - 2 - 3 475 180
AY451390.1 - 2 - 3 475 180
BC131517.1 - 3 - 4 498 180
HQ692852.1 - 1 - 2 400 180

View Full Product Details