The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182894.2 | NP_878314.1 | 2 - 3 | 540 | 116 |
GenBank mRNA | AY336059.1 | - | 2 - 3 | 540 | 116 |
BC128153.1 | - | 2 - 3 | 496 | 116 |