Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BARHL2
Entrez Gene ID
343472
Gene Name
BarH like homeobox 2
Gene Aliases
-
Chromosome Location
Chr.1: 90712022 - 90717237 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020063.1 NP_064447.1 3 - 3 970 143
GenBank mRNA AY338397.1 - 3 - 3 970 143
BC126439.1 - 3 - 3 954 143
BC126441.1 - 3 - 3 954 143

View Full Product Details