The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037329.3 | NP_001032406.1 | 3 - 4 | 384 | 102 |
XM_017017217.1 | XP_016872706.1 | 3 - 4 | 384 | 102 | |
XM_017017218.1 | XP_016872707.1 | 3 - 4 | 291 | 102 | |
XM_017017219.1 | XP_016872708.1 | 3 - 4 | 292 | 102 | |
GenBank mRNA | AK122736.1 | - | 3 - 4 | 377 | 102 |
BC040277.1 | - | 3 - 4 | 323 | 102 | |
BC106935.2 | - | 3 - 4 | 102 | ||
BC106936.2 | - | 3 - 4 | 102 | ||
DB238337.1 | - | 3 - 4 | 384 | 102 |