Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GBP6
Entrez Gene ID
163351
Gene Name
guanylate binding protein family member 6
Gene Aliases
-
Chromosome Location
Chr.1: 89363877 - 89388160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198460.2 NP_940862.2 2 - 3 463 109
XM_011540835.2 XP_011539137.1 2 - 3 292 109
GenBank mRNA AK131356.1 - 2 - 3 447 109
AK290303.1 - 2 - 3 283 109
AK315834.1 - 2 - 3 281 109
BC131713.1 - 2 - 3 241 109
BX537949.1 - 2 - 3 282 109
BX647907.1 - 2 - 3 280 109
DA437660.1 - 2 - 3 461 109
DA447005.1 - 2 - 3 447 109

View Full Product Details