The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153213.3 | NP_694945.2 | 15 - 16 | 2387 | 180 |
XM_011540706.2 | XP_011539008.1 | 16 - 17 | 2551 | 180 | |
GenBank mRNA | AK297707.1 | - | 11 - 12 | 1886 | 180 |
AL137736.1 | - | 10 - 11 | 1143 | 180 | |
BC012982.2 | - | 14 - 15 | 1963 | 180 | |
BC040640.1 | - | 15 - 16 | 2387 | 180 |