The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080532.1 | NP_001074001.1 | 15 - 16 | 1714 | 177 |
GenBank mRNA | AY236490.1 | - | 15 - 16 | 1792 | 177 |
AY263163.1 | - | 15 - 16 | 1753 | 177 | |
BC132679.1 | - | 15 - 16 | 1751 | 177 | |
BC143410.1 | - | 15 - 16 | 1751 | 177 | |
BC157874.1 | - | 15 - 16 | 1763 | 177 |