The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143763.1 | NP_001137235.1 | 8 - 9 | 630 | 165 |
NM_001143764.1 | NP_001137236.1 | 8 - 9 | 630 | 165 | |
NM_130784.2 | NP_570140.1 | 8 - 9 | 553 | 165 | |
GenBank mRNA | AK314978.1 | - | 8 - 9 | 580 | 165 |
AY027807.1 | - | 8 - 9 | 553 | 165 | |
AY027808.1 | - | 5 - 6 | 1489 | 165 | |
BC034821.1 | - | 8 - 9 | 580 | 165 |