Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBR4
Entrez Gene ID
23352
Gene Name
ubiquitin protein ligase E3 component n-recognin 4
Gene Aliases
RBAF600, ZUBR1, p600
Chromosome Location
Chr.1: 19074506 - 19210334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020765.2 NP_065816.2 75 - 76 11209 156
XM_011541108.2 XP_011539410.1 77 - 78 11444 156
XM_011541109.2 XP_011539411.1 77 - 78 11441 156
XM_011541110.2 XP_011539412.1 77 - 78 11441 156
XM_011541111.2 XP_011539413.1 77 - 78 11441 156
XM_011541112.2 XP_011539414.1 77 - 78 11429 156
XM_011541113.2 XP_011539415.1 77 - 78 11426 156
XM_011541114.2 XP_011539416.1 77 - 78 11426 156
XM_011541115.2 XP_011539417.1 77 - 78 11420 156
XM_011541116.2 XP_011539418.1 76 - 77 11411 156
XM_011541117.2 XP_011539419.1 76 - 77 11360 156
XM_011541118.2 XP_011539420.1 76 - 77 11357 156
XM_011541119.2 XP_011539421.1 77 - 78 11444 156
XM_011541120.2 XP_011539422.1 76 - 77 11321 156
XM_011541121.2 XP_011539423.1 75 - 76 11288 156
XM_017000822.1 XP_016856311.1 77 - 78 11423 156
XM_017000823.1 XP_016856312.1 76 - 77 11396 156
XM_017000824.1 XP_016856313.1 76 - 77 11342 156
XM_017000825.1 XP_016856314.1 75 - 76 11327 156
XM_017000826.1 XP_016856315.1 76 - 77 11324 156
XM_017000827.1 XP_016856316.1 75 - 76 11309 156
XM_017000828.1 XP_016856317.1 75 - 76 11285 156
XM_017000829.1 XP_016856318.1 75 - 76 11237 156
XM_017000830.1 XP_016856319.1 74 - 75 11186 156
GenBank mRNA AB007931.1 - 16 - 17 2485 156
AF348492.1 - 75 - 76 11205 156
AK023826.1 - 13 - 14 2040 155

View Full Product Details