Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- UBR4
- Entrez Gene ID
-
23352
- Gene Name
- ubiquitin protein ligase E3 component n-recognin 4
- Gene Aliases
-
RBAF600, ZUBR1, p600
- Chromosome Location
-
Chr.1: 19074506 - 19210334 on Build GRCh38
View Full Product Details