The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012486.2 | NP_036618.2 | 9 - 10 | 1315 | 135 |
XM_017001836.1 | XP_016857325.1 | 8 - 9 | 1128 | 135 | |
GenBank mRNA | AK292299.1 | - | 10 - 11 | 1267 | 135 |
U34349.1 | - | 7 - 8 | 979 | 135 |