Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR92
Entrez Gene ID
116143
Gene Name
WD repeat domain 92
Gene Aliases
-
Chromosome Location
Chr.2: 68130149 - 68157560 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256476.1 NP_001243405.1 3 - 4 532 122
NM_138458.3 NP_612467.1 3 - 4 532 122
NR_046234.1 3 - 4 405 122
GenBank mRNA AK056303.1 - 3 - 4 496 122
AK058090.1 - 3 - 4 405 122
AL834142.1 - 3 - 4 368 122
AY541587.1 - 3 - 4 507 122
BC014022.2 - 3 - 4 454 122
BC020271.1 - 3 - 4 486 122
BC036904.1 - 3 - 4 342 122
BC066657.1 - 3 - 4 482 122

View Full Product Details