Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFSD2B
Entrez Gene ID
388931
Gene Name
major facilitator superfamily domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.2: 24009022 - 24026775 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005264301.4 XP_005264358.1 13 - 14 2561 178
XM_017004063.1 XP_016859552.1 12 - 13 2654 178
XM_017004064.1 XP_016859553.1 12 - 13 2639 178
XM_017004065.1 XP_016859554.1 11 - 12 2570 178
GenBank mRNA BX647146.1 - 7 - 8 1265 178

View Full Product Details