Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO10
Entrez Gene ID
4651
Gene Name
myosin X
Gene Aliases
-
Chromosome Location
Chr.5: 16661907 - 16936276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012334.2 NP_036466.2 24 - 25 3025 180
XM_005248306.4 XP_005248363.1 6 - 7 1919 180
XM_005248307.2 XP_005248364.1 6 - 7 1395 180
XM_006714475.2 XP_006714538.1 23 - 24 2732 180
XM_011514046.2 XP_011512348.1 6 - 7 1098 180
GenBank mRNA AB018342.2 - 24 - 25 3012 180
AF132021.1 - 24 - 25 2637 180
AF234532.1 - 24 - 25 2779 180
AF247457.2 - 24 - 25 2710 180
AK295736.1 - 6 - 7 880 180
AK316138.1 - 6 - 7 1053 180
AL832428.1 - 14 - 15 1482 180
BC137168.1 - 24 - 25 2734 180
BC150285.1 - 24 - 25 3012 180

View Full Product Details