The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012334.2 | NP_036466.2 | 28 - 29 | 4365 | 134 |
XM_005248306.4 | XP_005248363.1 | 10 - 11 | 3259 | 134 | |
XM_005248307.2 | XP_005248364.1 | 10 - 11 | 2735 | 134 | |
XM_006714475.2 | XP_006714538.1 | 27 - 28 | 4072 | 134 | |
XM_011514046.2 | XP_011512348.1 | 10 - 11 | 2438 | 134 | |
GenBank mRNA | AB018342.2 | - | 28 - 29 | 4352 | 134 |
AF132021.1 | - | 28 - 29 | 3977 | 134 | |
AF234532.1 | - | 28 - 29 | 4119 | 134 | |
AF247457.2 | - | 28 - 29 | 4050 | 134 | |
AK074717.1 | - | 4 - 5 | 711 | 134 | |
AK295630.1 | - | 3 - 4 | 613 | 134 | |
AK295736.1 | - | 10 - 11 | 2220 | 134 | |
AK316138.1 | - | 10 - 11 | 2393 | 134 | |
AL832428.1 | - | 18 - 19 | 2822 | 134 | |
BC137168.1 | - | 28 - 29 | 4074 | 134 | |
BC150285.1 | - | 28 - 29 | 4352 | 134 |