The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012334.2 | NP_036466.2 | 29 - 30 | 4459 | 180 |
XM_005248306.4 | XP_005248363.1 | 11 - 12 | 3353 | 180 | |
XM_005248307.2 | XP_005248364.1 | 11 - 12 | 2829 | 180 | |
XM_006714475.2 | XP_006714538.1 | 28 - 29 | 4166 | 180 | |
XM_011514046.2 | XP_011512348.1 | 11 - 12 | 2532 | 180 | |
GenBank mRNA | AB018342.2 | - | 29 - 30 | 4446 | 180 |
AF132021.1 | - | 29 - 30 | 4071 | 180 | |
AF234532.1 | - | 29 - 30 | 4213 | 180 | |
AF247457.2 | - | 29 - 30 | 4144 | 180 | |
AK074717.1 | - | 5 - 6 | 805 | 180 | |
AK295630.1 | - | 4 - 5 | 707 | 180 | |
AK295736.1 | - | 11 - 12 | 2314 | 180 | |
AK316138.1 | - | 11 - 12 | 2487 | 180 | |
BC137168.1 | - | 29 - 30 | 4168 | 180 | |
BC150285.1 | - | 29 - 30 | 4446 | 180 |