The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003816.2 | NP_003807.1 | 19 - 20 | 2292 | 136 |
NR_027639.1 | 17 - 18 | 2105 | 136 | ||
NR_027878.1 | 18 - 19 | 2186 | 136 | ||
XM_011544682.2 | XP_011542984.1 | 19 - 20 | 2542 | 136 | |
XM_017013942.1 | XP_016869431.1 | 19 - 20 | 2235 | 136 | |
GenBank mRNA | AF495383.1 | - | 18 - 19 | 2141 | 136 |
AK293257.1 | - | 6 - 7 | 658 | 136 | |
BC143923.1 | - | 18 - 19 | 2146 | 136 | |
BC143924.1 | - | 17 - 18 | 2065 | 136 | |
D14665.2 | - | 15 - 16 | 1867 | 136 | |
U41766.1 | - | 19 - 20 | 2292 | 136 |