Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CLASP2
- Entrez Gene ID
-
23122
- Gene Name
- cytoplasmic linker associated protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 33496245 - 33718356 on Build GRCh38
View Full Product Details