Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLASP2
Entrez Gene ID
23122
Gene Name
cytoplasmic linker associated protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 33496245 - 33718356 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207044.1 NP_001193973.1 3 - 4 692 139
NM_015097.2 NP_055912.2 9 - 10 1153 139
XM_006713040.1 XP_006713103.1 9 - 10 1296 139
XM_006713041.1 XP_006713104.1 9 - 10 1293 139
XM_006713042.1 XP_006713105.1 9 - 10 1296 139
XM_006713048.1 XP_006713111.1 9 - 10 1296 139
XM_006713049.1 XP_006713112.1 9 - 10 1293 139
XM_006713050.1 XP_006713113.1 9 - 10 1296 139
XM_006713052.1 XP_006713115.1 9 - 10 1293 139
XM_006713053.1 XP_006713116.1 9 - 10 1296 139
XM_011533515.1 XP_011531817.1 9 - 10 1296 139
XM_017005947.1 XP_016861436.1 9 - 10 1293 139
XM_017005948.1 XP_016861437.1 9 - 10 1293 139
XM_017005949.1 XP_016861438.1 9 - 10 1139 139
XM_017005950.1 XP_016861439.1 9 - 10 1296 139
XM_017005951.1 XP_016861440.1 9 - 10 1293 139
XM_017005952.1 XP_016861441.1 9 - 10 1296 139
XM_017005953.1 XP_016861442.1 9 - 10 1293 139
XM_017005954.1 XP_016861443.1 9 - 10 1293 139
XM_017005955.1 XP_016861444.1 9 - 10 1296 139
XM_017005956.1 XP_016861445.1 9 - 10 1296 139
XM_017005957.1 XP_016861446.1 9 - 10 1296 139
XM_017005958.1 XP_016861447.1 9 - 10 1296 139
XM_017005959.1 XP_016861448.1 9 - 10 1293 139
XM_017005960.1 XP_016861449.1 9 - 10 1296 139
XM_017005961.1 XP_016861450.1 9 - 10 1139 139
XM_017005962.1 XP_016861451.1 9 - 10 1293 139
XM_017005963.1 XP_016861452.1 9 - 10 1296 139
XM_017005964.1 XP_016861453.1 9 - 10 1293 139
XM_017005965.1 XP_016861454.1 9 - 10 1296 139
XM_017005966.1 XP_016861455.1 9 - 10 1296 139
XM_017005967.1 XP_016861456.1 9 - 10 1139 139
XM_017005968.1 XP_016861457.1 3 - 4 436 139
XM_017005969.1 XP_016861458.1 3 - 4 446 139
XM_017005970.1 XP_016861459.1 3 - 4 695 139
XM_017005971.1 XP_016861460.1 3 - 4 446 139
XM_017005972.1 XP_016861461.1 3 - 4 1023 139
XM_017005973.1 XP_016861462.1 3 - 4 692 139
XM_017005974.1 XP_016861463.1 3 - 4 692 139
XM_017005975.1 XP_016861464.1 3 - 4 436 139
XM_017005976.1 XP_016861465.1 3 - 4 446 139
XM_017005977.1 XP_016861466.1 3 - 4 695 139
XM_017005978.1 XP_016861467.1 3 - 4 436 139
XM_017005979.1 XP_016861468.1 3 - 4 446 139
XM_017005980.1 XP_016861469.1 3 - 4 1023 139
XM_017005981.1 XP_016861470.1 3 - 4 436 139
XM_017005982.1 XP_016861471.1 3 - 4 446 139
XM_017005983.1 XP_016861472.1 3 - 4 695 139
XM_017005984.1 XP_016861473.1 3 - 4 692 139
XM_017005985.1 XP_016861474.1 3 - 4 1023 139
XM_017005986.1 XP_016861475.1 3 - 4 695 139
GenBank mRNA AB014527.2 - 3 - 4 331 139
AK090716.1 - 3 - 4 415 139
AK091451.1 - 9 - 10 1293 139
AK294345.1 - 3 - 4 483 139
BC029035.1 - 3 - 4 402 139
BC140778.1 - 9 - 10 989 139

View Full Product Details