The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126128.1 | NP_001119600.1 | 1 - 2 | 249 | 102 |
NM_021935.3 | NP_068754.1 | 1 - 2 | 249 | 102 | |
GenBank mRNA | AF182069.2 | - | 1 - 2 | 80 | 102 |
AF333025.1 | - | 1 - 2 | 104 | 102 | |
AY349131.1 | - | 1 - 2 | 95 | 102 | |
BC069395.1 | - | 1 - 2 | 104 | 102 | |
BC096695.1 | - | 1 - 2 | 104 | 102 | |
BC098110.1 | - | 1 - 2 | 104 | 102 | |
BC098162.1 | - | 1 - 2 | 104 | 102 |