The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173602.2 | NP_775873.2 | 5 - 6 | 795 | 83 |
XM_005269044.1 | XP_005269101.1 | 5 - 6 | 795 | 83 | |
XM_006719520.3 | XP_006719583.1 | 6 - 7 | 771 | 83 | |
XM_011538594.2 | XP_011536896.1 | 6 - 7 | 821 | 83 | |
XM_011538595.2 | XP_011536897.1 | 6 - 7 | 821 | 83 | |
XM_017019699.1 | XP_016875188.1 | 6 - 7 | 821 | 83 | |
XM_017019700.1 | XP_016875189.1 | 5 - 6 | 795 | 83 | |
XM_017019701.1 | XP_016875190.1 | 5 - 6 | 795 | 83 | |
XM_017019702.1 | XP_016875191.1 | 5 - 6 | 795 | 83 | |
GenBank mRNA | BC075027.2 | - | 5 - 6 | 673 | 83 |
BX537905.1 | - | 4 - 5 | 416 | 83 |