Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHISA5
Entrez Gene ID
51246
Gene Name
shisa family member 5
Gene Aliases
SCOTIN
Chromosome Location
Chr.3: 48467798 - 48504826 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011533796.1 XP_011532098.1 1 - 2 126 122

View Full Product Details