The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003214.3 | NP_003205.2 | 1 - 2 | 189 | 177 |
GenBank mRNA | AF142482.1 | - | 1 - 2 | 143 | 177 |
DA856821.1 | - | 1 - 2 | 212 | 177 |