The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243738.1 | NP_001230667.1 | 4 - 5 | 832 | 147 |
NM_004761.4 | NP_004752.1 | 5 - 6 | 1029 | 147 | |
XM_011514781.1 | XP_011513083.1 | 3 - 4 | 301 | 147 | |
XM_011514785.2 | XP_011513087.1 | 3 - 4 | 301 | 147 | |
GenBank mRNA | AB012295.1 | - | 4 - 5 | 381 | 147 |
AK294442.1 | - | 4 - 5 | 368 | 147 | |
AL050259.1 | - | 5 - 6 | 602 | 147 | |
BC005037.1 | - | 2 - 3 | 70 | 147 | |
BC032681.1 | - | 5 - 6 | 584 | 147 | |
BX648015.1 | - | 4 - 5 | 374 | 147 |