The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015692.2 | NP_056507.2 | 27 - 28 | 3806 | 156 |
XM_011527917.1 | XP_011526219.1 | 27 - 28 | 3773 | 156 | |
XM_011527918.1 | XP_011526220.1 | 27 - 28 | 3809 | 156 | |
XM_011527919.1 | XP_011526221.1 | 27 - 28 | 3809 | 156 | |
XM_011527920.2 | XP_011526222.1 | 27 - 28 | 3809 | 156 | |
XM_011527921.2 | XP_011526223.1 | 27 - 28 | 3809 | 156 | |
XM_011527922.1 | XP_011526224.1 | 27 - 28 | 3809 | 156 | |
XM_011527923.1 | XP_011526225.1 | 27 - 28 | 3809 | 156 | |
XM_011527924.2 | XP_011526226.1 | 27 - 28 | 3809 | 156 | |
XM_011527925.1 | XP_011526227.1 | 11 - 12 | 1651 | 156 | |
XM_017026594.1 | XP_016882083.1 | 27 - 28 | 3809 | 156 | |
GenBank mRNA | AB033109.1 | - | 27 - 28 | 3631 | 156 |
AY101765.1 | - | 27 - 28 | 3650 | 156 |