Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK024559.1 - 1 610 Chr. 7 - 48838899 - 48840345 [-] on Build GRCh38 152


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC105375273 LOC105375273 -127.4kb Anti-sense Chr. - 48708404 - 48711484 on Build GRCh38
Downstream CDC14 cell division cycle 14 C-like CDC14BL 6.1kb Sense Chr. - 48846402 - 48848081 on Build GRCh38

View Full Product Details