The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032246.4 | NP_115622.2 | 2 - 2 | 2961 | 121 |
GenBank mRNA | AB095929.1 | - | 1 - 1 | 3773 | 121 |
AK094145.1 | - | 1 - 1 | 2194 | 121 | |
AK094633.1 | - | 1 - 1 | 1625 | 121 | |
AK131424.1 | - | 2 - 2 | 2838 | 121 |