The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177540.2 | NP_803884.1 | 2 - 3 | 589 | 141 |
XM_017002403.1 | XP_016857892.1 | 1 - 2 | 1141 | 141 | |
GenBank mRNA | AF387619.1 | - | 1 - 2 | 270 | 141 |
AF387620.1 | - | 1 - 2 | 270 | 141 | |
AK292014.1 | - | 2 - 3 | 558 | 141 | |
AK293499.1 | - | 2 - 3 | 458 | 141 |