The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032482.2 | NP_115871.1 | 27 - 28 | 4645 | 116 |
GenBank mRNA | AF509504.1 | - | 27 - 28 | 4645 | 116 |
AK074120.1 | - | 23 - 24 | 4310 | 116 | |
BC021037.1 | - | 4 - 5 | 1362 | 116 |