The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146344.2 | NP_001139816.2 | 2 - 3 | 365 | 135 |
XM_011541479.2 | XP_011539781.1 | 1 - 2 | 551 | 135 | |
GenBank mRNA | AL049680.1 | - | 2 - 3 | 364 | 135 |
BC144426.1 | - | 2 - 3 | 364 | 135 |