The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173566.2 | NP_775837.2 | 8 - 9 | 6369 | 154 |
XM_006724219.3 | XP_006724282.1 | 8 - 9 | 6929 | 154 | |
XM_006724220.3 | XP_006724283.1 | 8 - 9 | 6331 | 154 | |
XM_006724221.2 | XP_006724284.1 | 7 - 8 | 5885 | 154 | |
XM_011530080.2 | XP_011528382.1 | 9 - 10 | 7137 | 154 | |
XM_011530081.2 | XP_011528383.1 | 8 - 9 | 7013 | 154 | |
XM_017028736.1 | XP_016884225.1 | 8 - 9 | 8089 | 154 | |
XM_017028737.1 | XP_016884226.1 | 7 - 8 | 7527 | 154 | |
GenBank mRNA | AK123082.1 | - | 3 - 4 | 345 | 154 |
BX647272.1 | - | 5 - 6 | 672 | 154 | |
CT841517.1 | - | 7 - 8 | 6201 | 154 |