The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129998.2 | NP_001123470.1 | 1 - 2 | 225 | 108 |
NM_001319241.1 | NP_001306170.1 | 1 - 2 | 225 | 108 | |
NM_001319242.1 | NP_001306171.1 | 1 - 2 | 225 | 108 | |
NM_205852.3 | NP_995324.2 | 1 - 2 | 225 | 108 | |
NR_135049.1 | 1 - 2 | 225 | 108 | ||
XM_011520658.2 | XP_011518960.1 | 2 - 3 | 236 | 108 | |
XM_011520663.2 | XP_011518965.1 | 2 - 3 | 316 | 108 | |
XM_017019295.1 | XP_016874784.1 | 2 - 3 | 305 | 108 | |
GenBank mRNA | AK128243.1 | - | 1 - 2 | 220 | 108 |
AK131027.1 | - | 1 - 2 | 93 | 108 | |
AY358810.1 | - | 1 - 2 | 225 | 108 | |
BC036036.1 | - | 1 - 2 | 93 | 108 | |
BC136805.1 | - | 1 - 2 | 165 | 108 | |
BC136806.1 | - | 1 - 2 | 165 | 108 | |
BG484230.1 | - | - | 498 | 108 | |
BM563861.1 | - | - | 92 | 108 | |
DQ368812.1 | - | 1 - 2 | 92 | 108 |