The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005160.2 | NP_001005160.1 | 1 - 1 | 839 | 150 |
GenBank mRNA | AY726590.1 | - | 1 - 1 | 775 | 150 |