Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GALT
Entrez Gene ID
2592
Gene Name
galactose-1-phosphate uridylyltransferase
Gene Aliases
-
Chromosome Location
Chr.9: 34646589 - 34650598 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000155.3 NP_000146.2 8 - 9 934 85
NM_001258332.1 NP_001245261.1 6 - 7 809 85
GenBank mRNA AK096026.1 - 4 - 5 898 85
AK096137.1 - 6 - 7 1278 85
AK303279.1 - 6 - 7 762 85
AK314340.1 - 8 - 9 916 85
BC015045.2 - 8 - 9 859 85
BT009852.1 - 8 - 9 818 85
KU178089.1 - 8 - 9 818 85
KU178090.1 - 8 - 9 885 85
M60091.1 - 8 - 9 846 85

View Full Product Details