Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- UNC80
- Entrez Gene ID
-
285175
- Gene Name
- unc-80 homolog, NALCN activator
- Gene Aliases
-
C2orf21, UNC-80
- Chromosome Location
-
Chr.2: 209771925 - 209999300 on Build GRCh38
View Full Product Details