The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018353.4 | NP_060823.3 | 13 - 14 | 3465 | 79 |
XM_005267833.4 | XP_005267890.1 | 14 - 15 | 3512 | 79 | |
XM_011536923.1 | XP_011535225.1 | 10 - 11 | 1950 | 79 | |
XM_017021425.1 | XP_016876914.1 | 15 - 16 | 3493 | 79 | |
GenBank mRNA | AB067490.1 | - | 13 - 14 | 3466 | 79 |
BC065544.1 | - | 13 - 14 | 3256 | 79 |